Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP79A4P
- Entrez Gene ID
-
833581
- Gene Name
- cytochrome P450, family 79, subfamily A, polypeptide 4 pseudogene
- Gene Aliases
-
''cytochrome P450, AT5G35920, F14A1.7, F14A1_7, cytochrome P450, family 79, polypeptide 4 pseudogene, polypeptide 4 pseudogene'', subfamily A
- Chromosome Location
-
Chr.5: 14056437 - 14056904 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122983.1 |
NP_198441.1 |
1 - 1 |
269 |
122 |
GenBank mRNA |
DQ056697.1 |
- |
1 - 1 |
269 |
122 |
View Full Product Details