Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG25
Entrez Gene ID
834246
Gene Name
SET domain protein 25
Gene Aliases
ARABIDOPSIS TRITHORAX-RELATED7, AT5G42400, ATXR7, MDH9.9, MDH9_9, SET domain protein 25
Chromosome Location
Chr.5: 16953515 - 16961245 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344444.1 NP_001318731.1 10 - 11 3050 108
NM_001344445.1 NP_001330664.1 10 - 11 3052 108
NM_001344446.1 NP_001330665.1 10 - 11 3041 108
NM_001344447.1 NP_001330668.1 10 - 11 3049 108
NM_001344448.1 NP_001330669.1 10 - 11 3049 108
NM_001344449.1 NP_001330666.1 10 - 11 3050 108
NM_001344450.1 NP_001330667.1 9 - 10 2872 108
NM_001344451.1 NP_001330670.1 10 - 11 3048 108
GenBank mRNA EU014691.1 - 9 - 10 2841 108

View Full Product Details