Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RAN1
- Entrez Gene ID
-
834509
- Gene Name
- copper-exporting ATPase / responsive-to-antagonist 1 / copper-transporting ATPase (RAN1)
- Gene Aliases
-
AT5G44790, HMA7, K23L20.14, K23L20_14, RESPONSIVE-TO-ANTAGONIST 1
- Chromosome Location
-
Chr.5: 18075562 - 18080160 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123847.3 |
NP_199292.1 |
3 - 4 |
1696 |
86 |
GenBank mRNA |
AF082565.1 |
- |
3 - 4 |
1507 |
86 |
View Full Product Details