Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CHR4
- Entrez Gene ID
-
834510
- Gene Name
- chromatin remodeling 4
- Gene Aliases
-
AT5G44800, K23L20.15, K23L20_15, MI-2-LIKE, PICKLE RELATED 1, PKR1, chromatin remodeling 4
- Chromosome Location
-
Chr.5: 18083277 - 18093907 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123848.4 |
NP_199293.3 |
4 - 5 |
1917 |
64 |
View Full Product Details