Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPS4
- Entrez Gene ID
-
834561
- Gene Name
- Disease resistance protein (TIR-NBS-LRR class) family
- Gene Aliases
-
AT5G45250, DISEASE RESISTANCE PROTEIN RPS4, K9E15.1, K9E15_1, RESISTANT TO P. SYRINGAE 4
- Chromosome Location
-
Chr.5: 18321764 - 18326175 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123893.2 |
NP_199338.1 |
4 - 5 |
3005 |
76 |
View Full Product Details