Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAT1
Entrez Gene ID
834666
Gene Name
1
Gene Aliases
AT5G46240, AtKAT1, MPL12.2, MPL12_2, potassium channel in Arabidopsis thaliana 1
Chromosome Location
Chr.5: 18743510 - 18747406 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123993.3 NP_199436.1 6 - 7 2001 124
GenBank mRNA AK176505.1 - 6 - 7 1201 124
AK220637.1 - 6 - 7 1201 124
BT046198.1 - 6 - 7 1156 124
M86990.1 - 6 - 7 1175 124

View Full Product Details