Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCB7
- Entrez Gene ID
-
834697
- Gene Name
- P-glycoprotein 7
- Gene Aliases
-
AT5G46540, ATP-binding cassette B7, K11I1.13, K11I1_13, P-glycoprotein 7, P-glycoprotein 7, PGP7
- Chromosome Location
-
Chr.5: 18877090 - 18882462 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124024.2 |
NP_199466.1 |
9 - 10 |
2686 |
107 |
View Full Product Details