Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP21
Entrez Gene ID
834717
Gene Name
ubiquitin-specific protease 21
Gene Aliases
AT5G46740, MZA15.16, MZA15_16, ubiquitin-specific protease 21
Chromosome Location
Chr.5: 18965271 - 18968798 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344697.1 NP_001330462.1 6 - 7 1588 76
NM_124044.2 NP_568667.1 6 - 7 1654 76

View Full Product Details