Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GFA2
Entrez Gene ID
834854
Gene Name
gametophytic factor 2
Gene Aliases
AT5G48030, MDN11.11, MDN11_11, gametophytic factor 2
Chromosome Location
Chr.5: 19465788 - 19469939 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124177.4 NP_568690.1 17 - 18 1463 67
GenBank mRNA AK118588.1 - 17 - 18 1370 67

View Full Product Details