Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMC3
Entrez Gene ID
834917
Gene Name
structural maintenance of chromosome 3
Gene Aliases
ARABIDOPSIS THALIANA CHROMOSOME ASSOCIATED PROTEIN-C, ARABIDOPSIS THALIANA STRUCTURAL MAINTENANCE OF CHROMOSOME 4, AT5G48600, ATCAP-C, ATSMC3, ATSMC4, K15N18.7, K15N18_7, STRUCTURAL MAINTENANCE OF CHROMOSOMES 3, structural maintenance of chromosome 3
Chromosome Location
Chr.5: 19701728 - 19709342 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203563.2 NP_001190492.1 13 - 14 1886 105
NM_124236.3 NP_199671.1 13 - 14 1737 105

View Full Product Details