Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAB1
Entrez Gene ID
835157
Gene Name
Transketolase family protein
Gene Aliases
AT5G50850, K16E14.6, MACCI-BOU
Chromosome Location
Chr.5: 20689243 - 20693470 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124463.4 NP_199898.1 4 - 5 480 130
GenBank mRNA AY070728.1 - 4 - 5 315 130
BT000839.1 - 3 - 4 187 130

View Full Product Details