Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRR1
Entrez Gene ID
835286
Gene Name
Dihydrodipicolinate reductase, bacterial/plant
Gene Aliases
AT5G52100, MSG15.20, MSG15_20, chlororespiration reduction 1
Chromosome Location
Chr.5: 21170048 - 21172346 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124589.3 NP_200023.1 8 - 9 928 120
GenBank mRNA BT030092.1 - 8 - 9 773 120

View Full Product Details