Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BETA-OHASE 2
Entrez Gene ID
835334
Gene Name
beta-carotene hydroxylase 2
Gene Aliases
AT5G52570, B2, BCH2, BETA CAROTENOID HYDROXYLASE 2, CHY2, F6N7.5, F6N7_5, FERREDOXIN-DEPENDENT DIIRON OXYGENASE, beta-carotene hydroxylase 2
Chromosome Location
Chr.5: 21334793 - 21337126 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124636.4 NP_200070.1 2 - 3 669 86
GenBank mRNA AY074394.1 - 2 - 3 452 86
AY117225.1 - 2 - 3 430 86

View Full Product Details