Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATIM
Entrez Gene ID
835368
Gene Name
timeless family protein
Gene Aliases
AT5G52910, MXC20.14, MXC20_14, TIMELESS
Chromosome Location
Chr.5: 21457534 - 21463327 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124669.3 NP_200103.1 5 - 6 1131 89

View Full Product Details