Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLT1
Entrez Gene ID
835427
Gene Name
NADH-dependent glutamate synthase 1
Gene Aliases
AT5G53460, MYN8.7, MYN8_7, NADH-GOGAT, NADH-dependent glutamate synthase 1
Chromosome Location
Chr.5: 21700187 - 21710296 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203600.1 NP_001190529.1 7 - 8 1515 70
NM_001203601.1 NP_001190530.1 7 - 8 1515 70
NM_124725.5 NP_200158.2 7 - 8 1530 70

View Full Product Details