Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CUC2
- Entrez Gene ID
-
835478
- Gene Name
- NAC (No Apical Meristem) domain transcriptional regulator superfamily protein
- Gene Aliases
-
ANAC098, AT5G53950, ATCUC2, Arabidopsis NAC domain containing protein 98, CUP-SHAPED COTYLEDON 2, K19P17.12, K19P17_12
- Chromosome Location
-
Chr.5: 21901704 - 21903854 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124774.3 |
NP_200206.1 |
2 - 3 |
530 |
109 |
GenBank mRNA |
BT030026.1 |
- |
2 - 3 |
471 |
109 |
View Full Product Details