Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HVA22J
Entrez Gene ID
818175
Gene Name
HVA22-like protein J
Gene Aliases
AT2G36020, F11F19.7, F11F19_7, HVA22-like protein J
Chromosome Location
Chr.2: 15123120 - 15125588 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129161.3 NP_565832.1 4 - 5 392 87
GenBank mRNA DQ446602.1 - 2 - 3 122 87

View Full Product Details