Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRF3
Entrez Gene ID
818213
Gene Name
growth-regulating factor 3
Gene Aliases
AT2G36400, AtGRF3, F1O11.3, F1O11_3, growth-regulating factor 3
Chromosome Location
Chr.2: 15270088 - 15273115 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129197.4 NP_181181.1 2 - 3 903 95
GenBank mRNA AY102636.1 - 2 - 3 599 95
BT026472.1 - 2 - 3 405 95

View Full Product Details