Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRF3
Entrez Gene ID
818213
Gene Name
growth-regulating factor 3
Gene Aliases
AT2G36400, AtGRF3, F1O11.3, F1O11_3, growth-regulating factor 3
Chromosome Location
Chr.2: 15270088 - 15273115 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129197.4 NP_181181.1 3 - 4 1260 71
GenBank mRNA AY102636.1 - 3 - 4 956 71
BT026472.1 - 3 - 4 762 71

View Full Product Details