Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SWC2
Entrez Gene ID
818246
Gene Name
DNA binding protein SWC2
Gene Aliases
AT2G36740, ATSWC2, F13K3.14, F13K3_14
Chromosome Location
Chr.2: 15406622 - 15409912 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336608.1 NP_001324358.1 2 - 3 242 141
NM_129229.4 NP_181212.2 2 - 3 319 141

View Full Product Details