Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXP3
Entrez Gene ID
818341
Gene Name
Barwin-like endoglucanases superfamily protein
Gene Aliases
ARABIDOPSIS THALIANA EXPANSIN A3, AT2G37640, ATEXP3, ATEXPA3, ATHEXP ALPHA 1.9, EXPANSIN 3, F13M22.14, F13M22_14
Chromosome Location
Chr.2: 15787523 - 15790139 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336666.1 NP_001324717.1 1 - 2 429 124
NM_129320.4 NP_181300.1 2 - 3 803 124
GenBank mRNA AY090368.1 - 2 - 3 517 124
AY122898.1 - 2 - 3 476 124

View Full Product Details