Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATATH8
Entrez Gene ID
818504
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT2G39190, T16B24.17, T16B24_17
Chromosome Location
Chr.2: 16349968 - 16356097 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129476.3 NP_565900.1 10 - 11 1709 139
NM_129477.1 NP_565901.1 10 - 11 1705 139
GenBank mRNA AY048299.1 - 10 - 11 1705 139

View Full Product Details