Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATH9
Entrez Gene ID
818597
Gene Name
ABC2 homolog 9
Gene Aliases
ABC2 homolog 9, AT2G40090, ATATH9, F27I1.1, F27I1_1
Chromosome Location
Chr.2: 16737466 - 16740486 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129567.4 NP_565923.1 1 - 2 361 70
GenBank mRNA AK175658.1 - 1 - 2 191 70
AK176037.1 - 1 - 2 191 70

View Full Product Details