Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK20
Entrez Gene ID
818888
Gene Name
MAP kinase 20
Gene Aliases
AT2G42880, ATMPK20, F7D19.12, F7D19_12, MAP kinase 20
Chromosome Location
Chr.2: 17840168 - 17844380 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336999.1 NP_001324574.1 2 - 3 863 142
NM_129849.5 NP_565989.1 2 - 3 863 142
GenBank mRNA AF412082.1 - 2 - 3 726 142
BT001021.1 - 2 - 3 430 142

View Full Product Details