Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUM2
Entrez Gene ID
817468
Gene Name
pumilio 2
Gene Aliases
APUM2, AT2G29190, F16P2.43, F16P2_43, pumilio 2
Chromosome Location
Chr.2: 12543812 - 12548563 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202701.1 NP_001189630.1 9 - 10 3257 160
NM_128475.4 NP_180482.1 9 - 10 3239 160

View Full Product Details