Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOT5
Entrez Gene ID
837889
Gene Name
C2H2-like zinc finger protein
Gene Aliases
AT1G13290, DEFECTIVELY ORGANIZED TRIBUTARIES 5, T6J4.5, T6J4_5, WIP domain protein 6, WIP6
Chromosome Location
Chr.1: 4550153 - 4551620 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101200.2 NP_172787.1 1 - 2 437 139
GenBank mRNA AJ311809.1 - 1 - 2 437 139

View Full Product Details