Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPX1
Entrez Gene ID
842910
Gene Name
thioredoxin-dependent peroxidase 1
Gene Aliases
AT1G65980, F12P19.14, F12P19_14, PEROXIREDOXIN TPX1, thioredoxin-dependent peroxidase 1
Chromosome Location
Chr.1: 24559253 - 24560980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105270.3 NP_176773.1 2 - 3 464 114
GenBank mRNA AF121355.1 - 2 - 3 290 114
AF332464.1 - 2 - 3 237 114
AY065056.1 - 2 - 3 300 114

View Full Product Details