Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAMMA CAL2
Entrez Gene ID
824029
Gene Name
gamma carbonic anhydrase-like 2
Gene Aliases
AT3G48680, AtCAL2, gamma carbonic anhydrase-like 2
Chromosome Location
Chr.3: 18034935 - 18037041 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114727.4 NP_190437.1 1 - 2 565 136
GenBank mRNA AF360214.1 - 1 - 2 489 136
AY051017.1 - 1 - 2 393 136

View Full Product Details