Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71B12
- Entrez Gene ID
-
832584
- Gene Name
- cytochrome P450, family 71, subfamily B, polypeptide 12
- Gene Aliases
-
''cytochrome P450, AT5G25130, F21J6.2, cytochrome P450, family 71, polypeptide 12, polypeptide 12'', subfamily B
- Chromosome Location
-
Chr.5: 8668232 - 8670237 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122422.4 |
NP_197895.1 |
2 - 3 |
1009 |
140 |
GenBank mRNA |
BT030466.1 |
- |
1 - 2 |
874 |
140 |
View Full Product Details