Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SHN3
- Entrez Gene ID
-
832611
- Gene Name
- Integrase-type DNA-binding superfamily protein
- Gene Aliases
-
AT5G25390, F18G18.130, F18G18_130, shine2, shine3
- Chromosome Location
-
Chr.5: 8820458 - 8822062 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122448.4 |
NP_197921.1 |
1 - 2 |
253 |
116 |
View Full Product Details