Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILL1
Entrez Gene ID
835766
Gene Name
IAA-leucine resistant (ILR)-like 1
Gene Aliases
AT5G56650, IAA-leucine resistant (ILR)-like 1, MIK19.10, MIK19_10
Chromosome Location
Chr.5: 22930775 - 22932822 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345211.1 NP_001332665.1 3 - 4 848 122
NM_125048.2 NP_200476.1 3 - 4 848 122
GenBank mRNA AK176293.1 - 3 - 4 798 122
AK176320.1 - 3 - 4 798 122
BT033041.1 - 3 - 4 798 122
U23795.1 - 3 - 4 798 122

View Full Product Details