Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl39
Entrez Gene ID
824365
Gene Name
serine carboxypeptidase-like 39
Gene Aliases
AT3G52020, serine carboxypeptidase-like 39
Chromosome Location
Chr.3: 19299309 - 19301148 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115061.2 NP_190770.1 3 - 3 1119 81

View Full Product Details