Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBD11
Entrez Gene ID
817398
Gene Name
LOB domain-containing protein 11
Gene Aliases
AT2G28500, LOB domain-containing protein 11, T17D12.6, T17D12_6
Chromosome Location
Chr.2: 12186274 - 12188293 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128410.3 NP_180417.2 2 - 2 769 74

View Full Product Details