Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP70T-2
Entrez Gene ID
817771
Gene Name
heat-shock protein 70T-2
Gene Aliases
AT2G32120, F22D22.13, F22D22_13, heat-shock protein 70T-2
Chromosome Location
Chr.2: 13651442 - 13653945 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128771.4 NP_180771.1 2 - 2 1869 138
NM_179852.3 NP_850183.1 2 - 2 1856 138
GenBank mRNA AY123990.1 - 2 - 2 1735 138

View Full Product Details