Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GER1
- Entrez Gene ID
-
843659
- Gene Name
- GDP-4-keto-6-deoxymannose-3,5-epimerase-4-reductase 1
- Gene Aliases
-
''GDP-4-keto-6-deoxymannose-3, 5-EPIMERASE-4-REDUCTASE, 5-epimerase-4-reductase 1, 5-epimerase-4-reductase 1'', ACTIVATING TRANSCRIPTION FACTOR 5, AT1G73250, ATFX, GDP-4-KETO-6-DEOXY-D-MANNOSE-3, GDP-4-keto-6-deoxymannose-3, T18K17.8, T18K17_8
- Chromosome Location
-
Chr.1: 27544969 - 27546479 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105984.4 |
NP_177468.2 |
2 - 2 |
1190 |
148 |
View Full Product Details