The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_125810.3 | NP_201219.1 | 2 - 2 | 830 | 152 |
GenBank mRNA | AY059859.1 | - | 2 - 2 | 333 | 152 |
AY087546.1 | - | 2 - 2 | 346 | 152 | |
L09755.1 | - | 2 - 2 | 329 | 152 |