Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCRL7
Entrez Gene ID
3767593
Gene Name
SCR-like 7
Gene Aliases
AT1G60989, SCR-like 7
Chromosome Location
Chr.1: 22462160 - 22462721 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036137.3 NP_001031214.1 1 - 2 141 93
GenBank mRNA DQ912182.1 - 1 - 2 71 93

View Full Product Details