Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR34
Entrez Gene ID
3770009
Gene Name
low-molecular-weight cysteine-rich 34
Gene Aliases
AT4G09984, low-molecular-weight cysteine-rich 34
Chromosome Location
Chr.4: 6254210 - 6255550 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036530.2 NP_001031607.1 1 - 2 129 144

View Full Product Details