Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARPC4
Entrez Gene ID
3770404
Gene Name
binding protein
Gene Aliases
AT4G14147
Chromosome Location
Chr.4: 8153807 - 8156975 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036555.3 NP_001031632.1 5 - 6 352 132
NM_001203797.1 NP_001190726.1 6 - 7 385 132
NM_001340903.1 NP_001319932.1 6 - 7 570 132

View Full Product Details