Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSR3
Entrez Gene ID
815962
Gene Name
vaculolar sorting receptor 3
Gene Aliases
AT2G14740, ATVSR3, BP80-2;2, T6B13.4, VACUOLAR SORTING RECEPTOR 2;2, VACUOLAR SORTING RECEPTOR 3, VSR2;2, binding protein of 80 kDa 2;2, vaculolar sorting receptor 3
Chromosome Location
Chr.2: 6308683 - 6312726 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127038.2 NP_179081.1 3 - 4 1302 82
NM_179624.2 NP_849955.1 3 - 4 1302 82

View Full Product Details