Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLS
Entrez Gene ID
3770598
Gene Name
polari
Gene Aliases
AT4G39403, POLARIS
Chromosome Location
Chr.4: 18329094 - 18329699 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036736.1 NP_001031813.1 1 - 1 551 177
GenBank mRNA AF285768.1 - 1 - 1 551 177

View Full Product Details