Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIF3
Entrez Gene ID
5007704
Gene Name
mini zinc finger
Gene Aliases
AT1G18835, mini zinc finger, mini zinc finger 3
Chromosome Location
Chr.1: 6495844 - 6496491 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084091.2 NP_001077560.1 1 - 1 247 73
GenBank mRNA DQ217608.1 - 1 - 1 152 73

View Full Product Details