Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOB1A
Entrez Gene ID
614611
Gene Name
MOB kinase activator 1A
Gene Aliases
MOBKL1B
Chromosome Location
Chr.11: 10475458 - 10498036 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076338.1 NP_001069806.1 2 - 3 364 104
GenBank mRNA BC120136.1 - 2 - 3 364 104

View Full Product Details