Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHPS
Entrez Gene ID
444988
Gene Name
deoxyhypusine synthase
Gene Aliases
DYS
Chromosome Location
Chr.7: 12816113 - 12820023 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003657.1 NP_001003657.1 1 - 2 538 56
GenBank mRNA AJ577610.2 - 1 - 2 538 56
BC105230.1 - 1 - 2 307 56

View Full Product Details