Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPN
Entrez Gene ID
507990
Gene Name
asporin
Gene Aliases
-
Chromosome Location
Chr.8: 84067280 - 84092633 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034309.2 NP_001029481.1 6 - 7 995 112
GenBank mRNA BC103199.1 - 6 - 7 996 112

View Full Product Details