Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPK2
Entrez Gene ID
281569
Gene Name
uroplakin 2
Gene Aliases
-
Chromosome Location
Chr.15: 29529296 - 29548145 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174214.2 NP_776639.1 4 - 5 458 90
GenBank mRNA L20633.1 - 4 - 5 458 90

View Full Product Details