Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP3
Entrez Gene ID
281758
Gene Name
fatty acid binding protein 3
Gene Aliases
FABP, FABP-3, H-FABP
Chromosome Location
Chr.2: 122287099 - 122294639 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174313.2 NP_776738.1 2 - 3 292 65
GenBank mRNA BC102153.1 - 2 - 3 307 65
BT021486.1 - 3 - 4 362 65
FJ756345.1 - 2 - 3 246 65
X12710.1 - - 292 65
X51933.1 - - 283 65

View Full Product Details