Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP3
Entrez Gene ID
281758
Gene Name
fatty acid binding protein 3
Gene Aliases
FABP, FABP-3, H-FABP
Chromosome Location
Chr.2: 122287099 - 122294639 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174313.2 NP_776738.1 3 - 4 393 114
GenBank mRNA BC102153.1 - 3 - 4 408 114
BT021486.1 - 4 - 5 463 114
X12710.1 - - 393 114
X51933.1 - - 384 114

View Full Product Details