Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR5A1
Entrez Gene ID
281948
Gene Name
nuclear receptor subfamily 5 group A member 1
Gene Aliases
SF-1
Chromosome Location
Chr.11: 95513838 - 95538616 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174403.2 NP_776828.1 5 - 6 1108 136
GenBank mRNA D13569.1 - 5 - 6 1108 136

View Full Product Details