Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPN1SW
Entrez Gene ID
282294
Gene Name
opsin 1, short wave sensitive
Gene Aliases
BCP
Chromosome Location
Chr.4: 92745939 - 92748928 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174567.1 NP_776992.1 4 - 5 937 107
GenBank mRNA U92557.1 - 4 - 5 937 107

View Full Product Details